Lab Jacks
Lab Jacks
Actual product may vary
Lab Jacks
Lab Jacks
Lab Jacks
Precise height adjustment with the turn of a knob.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Height (English)Capacity (English)Dimensions (L x W)
FISS6308315.2 to 50.8 cm3.5 to 20 in.100 lb.12 x 12 in. (Plate)
FISS630792.54 to 8.12 cm1 to 3.2 in.8.8 lb.3 x 3 in. (Plate)
FISS630825.7 to 27.3 cm2.25 to 11 in.79 lb.8 x 8 in. (Plate)
FISS630815.7 to 27.3 cm2.45 to 10.75 in.60 lb.6 x 6 in. (Plate)
Catalog number FISS63083
Price (SGD)
-
Height (Metric):
15.2 to 50.8 cm
Height (English):
3.5 to 20 in.
Capacity (English):
100 lb.
Dimensions (L x W):
12 x 12 in. (Plate)
  • For supporting hotplates, baths, glassware and other items that require stable and precise height regulation
  • Large diameter knobs provide a smooth and easy height adjustment
  • Platforms are made of nonsparking anodized aluminum
  • Standard, large and giant lab jacks have threaded holes to accommodate an optional support rod (S23514) and other apparatus
Specifications
MaterialAnodized Aluminum (Top), Stainless Steel
Height (Metric)15.2 to 50.8 cm
Height (English)3.5 to 20 in.
Dimensions (L x W)12 x 12 in. (Plate)
Capacity (English)100 lb.
Product TypeLab Jack
SizeGiant
For Use With (Equipment)Supporting hotplates, baths, glassware
Unit SizeEA