Kik-Step™ Stool
Kik-Step™ Stool
Actual product may vary
Kik-Step™ Stool
The Kik-Step™ Stool is a rolling stool with a rubber bumper to protect walls and equipment.
Have Questions?
Catalog NumberLength (English)
FISS5110114.5 in.
Catalog number FISS51101
Price (SGD)
-
Length (English):
14.5 in.
  • Step on it and spring-loaded casters retract for safe, sure footing
  • Step off and stool rolls smoothly
  • Two steel steps cushioned with rubber treads
  • Epoxy finish protected by wide rubber bumper around base
Specifications
ColorGray
Diameter (English) Base15.5 in.
Diameter (Metric) Base39.37 cm
FinishEpoxy
Height (English)14.5 in.
Height (Metric)36.83 cm
IncludesRubber matting and bumper, spring loaded casters
Length (English)14.5 in.
Weight (English)10 lb.
Weight (Metric)4.53 kg
Load Capacity (English)350 lb.
Load Capacity (Metric)158.75 kg
MaterialStainless Steel
Product TypeStep Stool
Unit SizeEA