Stainless Steel Micro Spatulas, Double-ended, tapered and rounded ends
Stainless Steel Micro Spatulas, Double-ended, tapered and rounded ends
Actual product may vary
Stainless Steel Micro Spatulas, Double-ended, tapered and rounded ends
Transfer small quantities from semimicro centrifuge and test tubes.
Have Questions?
Catalog NumberLength (Metric)
FISS5082220.32 cm
Catalog number FISS50822
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
20.32 cm
Handy tools to scrape solidified material from crucibles and jars.
Stock up on these stainless steel micro-spatulas.
  • Thin, flexible, rounded and tapered ends.
  • Convenient, multi-purpose design
  • High Quality Stainless Steel
  • Durable, and reliable for educational and research experiments
  • Tapered End: Designed to be used for labware with conical-bottom tubes – 0.14 in. (3.5mm)
  • Rounded End: Designed to be used for regular/test tubes – 0.25in (6.6mm)
Specifications
Product TypeMicro Spatula
For Use With (Equipment)Crucibles and jars
For Use With (Application)Designed to scrape solidified material from crucibles and jars
Length (English)8 in.
IncludesDouble ended with one tapered end and one rounded end
MaterialStainless Steel
Length (Metric)20.32 cm
Unit SizePK