Spoon and Spatula Tool
Spoon and Spatula Tool
Actual product may vary
Spoon and Spatula Tool
Versatile tool ideal for a wide range of applications.
Have Questions?
Catalog NumberLength (Metric)
FISS50789A22.9 cm
Catalog number FISS50789A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
22.9 cm
  • 9 in. (22.9 cm) long
  • Polished
  • Sold in packs of six
Specifications
AutoclavableAutoclavable
ShapeSpoon End, Spatula End
Product TypeSpoon Spatula
FinishPolished
For Use With (Application)Extremely versatile tool for a wide range of applications
Length (English)9 in.
IncludesSpoon and spatula ends
MaterialStainless Steel
Length (Metric)22.9 cm
Unit SizePK