Polypropylene Tubing Connectors and Adapters
Polypropylene Tubing Connectors and Adapters
Actual product may vary
Fisher Catalogue
Polypropylene Tubing Connectors and Adapters
Work in any tubing or vacuum system.
Have Questions?
Catalog NumberFor Use WithOverall Length (Metric)Shape
FISS507093 to 7.8mm83.82 mmStraight-stepped
Catalog number FISS50709
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With:
3 to 7.8mm
Overall Length (Metric):
83.82 mm
Shape:
Straight-stepped
  • Excellent chemical and temperature resistance
  • Serrated connecting surfaces to prevent tubing slippage
  • Autoclavable Polypropylene
  • Adapters allow for connecting different diameter tubing
Specifications
AutoclavableAutoclavable
Diameter (English) Inner0.125 to 0.312 in. to 0.438 to 0.625 in.
MaterialPolypropylene
Overall Length (English)3.3 in.
Overall Length (Metric)83.82 mm
For Use With3 to 7.8mm
For Use With (Application)Used for any tubing or vacuum system
Width (Metric)20.5 mm
Width (English)0.81 in.
ShapeStraight-stepped
ColorWhite
Maximum Pressure20 psi
Product TypeTubing Connector
Unit SizePK