Bench-Top Biohazard Bag Holder
Bench-Top Biohazard Bag Holder
Actual product may vary
Bel-Art
Bench-Top Biohazard Bag Holder
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
Have Questions?
Catalog NumberProduct Type
FISS50397ABiohazard Bag Holder
Catalog number FISS50397A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Biohazard Bag Holder
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28 cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812
Specifications
AutoclavableAutoclavable
ColorOrange
DisposableSingle-use
For Use With (Application)Perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
For Use With (Equipment)01-828A, 01-828-031, 01-830A, 01-826-1, 01-827A, 01-829A, 01-815 11 H x 8.5 in. W Biohazard Bags biohazard bag, Clavies autoclavable bags (01-815-6)
IncludesRectangular stainless steel wire frame only
MaterialEpoxy-coated Steel Wire
Dimensions (L x W x H)3.4 x 5 x 8.5 in. (8.63 x 12.7 x 21.59 cm)
Length (English)3.4 in.
Length (Metric)8.63 cm
Width (English)5 in.
Width (Metric)12.7 cm
Height (English)8.5 in.
Height (Metric)21.59 cm
Product TypeBiohazard Bag Holder
Unit SizeEA