Policemen Stirring Rod
Actual product may vary
Policemen Stirring Rod
Policemen Stirring Rod
Bel-Art
Policemen Stirring Rod
Round molded polypropylene rods.
Have Questions?
Catalog NumberLength (Metric)
FISS50357203 mm
Catalog number FISS50357
Price (SGD)
-
Length (Metric):
203 mm
  • One end flattened and cut diagonally
  • Dimensions: 0.18 dia. x 8 in. (4.67 x 203 mm)
  • Steam autoclavable at 121°C (250°F)
  • Sold in packs of six
Specifications
AutoclavableAutoclavable
Diameter (English)0.18 in.
For Use With (Application)For larger work areas
Length (English)8 in.
Diameter (Metric)4.67 mm
Length (Metric)203 mm
MaterialPolypropylene
Quantity6/Pk.
ShapeCylinder
TypeStirring Rod
Unit SizePack