Cork Borer Sets with Handles
Cork Borer Sets with Handles
Actual product may vary
Cork Borer Sets with Handles
Bore holes in corks and rubber stoppers.
Have Questions?
Catalog NumberDiameter (Metric) OuterDiameter (English) OuterQuantity
FISS50166A4.8 to 7.8mm0.18 to 0.31 in.3 Pack
Catalog number FISS50166A
Price (SGD)
-
Diameter (Metric) Outer:
4.8 to 7.8mm
Diameter (English) Outer:
0.18 to 0.31 in.
Quantity:
3 Pack
  • Consist of plated tubing with precision-ground cutting edges
  • Smooth plastic handles
  • Handles are stamped with a number designation
  • Punch included with each set
Specifications
Product TypeCork Borer
Diameter (English) Outer0.18 to 0.31 in.
Diameter (Metric) Outer4.8 to 7.8mm
For Use With (Application)To bore holes in corks and Rubber stoppers
IncludesPlated tubing, smooth Plastic handles
Quantity3 Pack
Unit SizePK