Screw-Compression Pinchcock
Screw-Compression Pinchcock
Actual product may vary
Fisher Catalogue
Screw-Compression Pinchcock
Open sided—easily fits over tubing to regulate air/liquid flow.
Have Questions?
Catalog number FISS49137
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Features a 1 in. (25mm) diameter serrated head for greater leverage
  • Constructed from brass and plated for corrosion resistance
Specifications
TypeScrew Compressor Clamp
MaterialBrass
Unit SizePK