Check Order Status
Silver Nitrate (Crystalline/USP), Fisher Chemical™
Element Search
Structure Search
Silver Nitrate (Crystalline/USP), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FISS486-100
100 g
Glass Bottle
Catalog number
FISS486-100
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Boiling Point
444°C
Chemical Name or Material
Silver Nitrate
Melting Point
212°C
CAS
7761-88-8
ChemAlert Storage Symbol
Yellow
View more