Tripod Base Support Stands
Actual product may vary
American Educational Products
Tripod Base Support Stands
Heavy-duty, tripod base support stands provide years of solid support in the laboratory.
Have Questions?
Catalog NumberLength (English) RodDiameter (English) Rod
FISS4780918 in.0.3 in.
Catalog number FISS47809
Price (SGD)
-
Length (English) Rod:
18 in.
Diameter (English) Rod:
0.3 in.
  • Available in various lengths and rod diameters
  • Feature threaded bases and threaded rods for extra stability
  • Bases constructed of rugged cast-iron and finished with chemical-resistant, baked black enamel for added stability and durability
Specifications
MaterialCast Iron
Length (English) Rod18 in.
Length (Metric) Rod48.3 cm
Diameter (English) Rod0.3 in.
Diameter (Metric) Rod0.8 cm
Product TypeTripod Base Support Stand
FinishBaked Black Enamel
For Use With (Application)Provide years of solid support in the laboratory
QuantityEach
Unit SizeEach