Visorgogs™ Goggles
Visorgogs™ Goggles
Actual product may vary
Visorgogs™ Goggles
Designed to protect eyes from flying particles or liquids.
Have Questions?
Catalog number FISS47619S
Price (SGD)
-
  • Offer complete protection from glaring overhead light
  • Lenses are one-piece, impact-resistant plastic with a down-angle vision
Specifications
Lens CoatingUncoated
Certifications/ComplianceANSI Z87.1-2003 (Z87+ High Impact).
Lens TintClear
Product TypeFull-view Goggles
Ventilation DetailsScreened
Unit SizeEA