Kimwipes™ Delicate Task Wipers
Kimwipes™ Delicate Task Wipers
Actual product may vary
Kimwipes™ Delicate Task Wipers
Clean and polish delicate surfaces without scratching or leaving residue. Single-ply Kimwipes™ EX-L wipes are made of a finely textured tissue. Come in a convenient Pop-U™ box with antistatic Lintguard™ polyshield.
Have Questions?
Catalog NumberDimensions (L x W)
FISS472998.4 x 4.4 in.
Catalog number FISS47299
Price (SGD)
-
Dimensions (L x W):
8.4 x 4.4 in.
  • Low-linting wipers absorb 18% more water and are 24% faster than the leading private label task wiper
  • Ideal for surfaces, parts, instruments and lens cleaning in laboratories and research environments
  • Will not scratch delicate surfaces when used wet
  • Dispensed individually to reduce waste while minimizing lint, static buildup, and contamination
  • Specifications
    Product TypeDelicate Task Wipe
    Length (English)4 in.
    Length (Metric)21.33 cm
    Width (English)8 in.
    Width (Metric)11.17 cm
    MaterialTissue
    ColorWhite
    DisposableSingle-use
    Dimensions (L x W)8.4 x 4.4 in.
    For Use With (Application)Ideal for surfaces, parts, instruments and lens cleaning in laboratories and research environment
    Ply1-ply
    Unit SizePK