Wall-Mount Clock, Calendar, and Thermometer
Wall-Mount Clock, Calendar, and Thermometer
Actual product may vary
Wall-Mount Clock, Calendar, and Thermometer
High-tech design looks great in any science classroom or office.
Have Questions?
Catalog number FISS43192
Price (SGD)
-
  • Read the time, date and temperature with one glance at the large 6.5 × 3.75 in. display
  • Time can be displayed in 12-hour AM/PM or 24-hour international time
  • Temperature is °C/°F selectable
  • Calendar displays month, date and day of the week
Specifications
Product TypeCalendar Thermometer Clock
Accuracy0.0001
Resolution1 sec.
DisplayLCD
Battery TypeAAA
Temperature Range (English)23°F to 122°F
Temperature Range (Metric)-5°C to +50°C
Temperature Resolution0.5°
Dimensions (L x W)9.25 x 10 in. (23.5 x 25.4 cm)
Length (English)9.25 in.
Length (Metric)23.5 cm
Width (English)10 in.
Width (Metric)25.4 cm
Depth (English)1 in.
Depth (Metric)2.54 cm
Weight (English)19 oz.
Weight (Metric)0.53 kg
IncludesThree AAA batteries, wall mount
For Use With (Application)High tech design looks great in any science classroom or office
QuantityEach
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEA