Neodymium Disc Magnets
Neodymium Disc Magnets
Actual product may vary
Neodymium Disc Magnets
For use in automobile and electronic devises.
Have Questions?
Catalog NumberGrade
FISS43047135
Catalog number FISS430471
Price (SGD)
-
Grade:
35
  • Composed of neodymium, iron, boron and a few transition metals
  • Resistance to demagnetization
  • Good in ambient temperature
  • Material is corrosive and should be coated for long term maximum energy output
  • Grade: 35
Magnetic separators, Linear actuators, Microphone assemblies, Computer rigid disc drives, printers and speakers
Specifications
Diameter (English) Outside1 in.
Diameter (Metric) Outside2.54 cm
Weight (English)0.15738 lb.
Weight (Metric)0.0714 kg
Grade35
Product TypeNeodymium Disc Magnet
For Use With (Application)Automobile and electronic devises,
Unit SizeEA