Check Order Status
Sodium Sulfide Hydrate (Flakes/Technical), Fisher Chemical
Element Search
Structure Search
Sodium Sulfide Hydrate (Flakes/Technical), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FISS426-212
2.5 kg
Glass Bottle
Catalog number
FISS426-212
Price (SGD)
-
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Sulfide
CAS
1313-82-2
ChemAlert Storage Symbol
White
Color
Yellow
CAS
7732-18-5
View more