Alcohol Burner
Alcohol Burner
Actual product may vary
Alcohol Burner
For use with any experiment requiring low heat.
Have Questions?
Catalog number FISS41898B
Price (SGD)
-
For use with any experiment requiring low heat.
  • Polished metal alcohol burner
  • Flame can be easily adjusted by rotating ratcheted wick control wheel
  • With air cap attached by matching polished chain
  • Safer than glass burner
  • Dimensions: 4.5 x 3 x 4.5 in. (11.43 x 7.62 x 11.43cm)
  • Capacity: 150mL
  • Weight: 0.5 lb. (0.22kg)
  • Replacement wicks also available (Cat. No. S05050)
Specifications
Product TypeAlcohol Burner
MaterialPolished Metal
Capacity (English)0.039 gal.
Capacity (Metric)150 mL
Diameter (English)2.75 in.
Diameter (Metric)6.4 cm
For Use With (Application)Experiments Requiring Low Heat
Height (English)4.5 in.
Height (Metric)11.43 cm
IncludesAir Cap
Weight (English)0.5 lb.
Weight (Metric)0.22 kg
Unit SizeEA