Plastic Weighing Scoops
Plastic Weighing Scoops
Actual product may vary
Fisherbrand
Plastic Weighing Scoops
Eisco™ Plastic Weighing Scoops are constructed of polypropylene and are the ideal replacement for metallic scoops.
Have Questions?
Catalog NumberCapacity (Metric)Quantity
FISS41657100 mL1
Catalog number FISS41657
Price (SGD)
-
Capacity (Metric):
100 mL
Quantity:
1
  • Plastic weigh scoop with handle
  • Made from polypropylene
  • Autoclavable
Specifications
Product TypeScoop
MaterialPolypropylene
AutoclavableAutoclavable
Capacity (English)3.38 oz.
Capacity (Metric)100 mL
DisposableReusable
SterilityNon-sterile
ColorWhite
For Use With (Application)Measuring chemicals
Quantity1
Height (Metric)3.8 cm
Length (Metric)20.32 cm
Width (Metric)7 cm
Unit SizeEA