SP Scienceware™ LabMat™ Bench Liners
Actual product may vary
Bel-Art
SP Scienceware™ LabMat™ Bench Liners
Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.
Have Questions?
Catalog NumberDimensions (L x W)ColorQuantity
FISS4126750 ft. x 20 in.White50 ft./Roll
Catalog number FISS41267
Price (SGD)
-
Dimensions (L x W):
50 ft. x 20 in.
Color:
White
Quantity:
50 ft./Roll
  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials
Specifications
Product TypeBench Protector
MaterialPolyethylene
Absorbancy420 mL/m2
Length (English)50 ft.
Length (Metric)15.2 m
Width (English)20 in.
Width (Metric)50.8 cm
Dimensions (L x W)50 ft. x 20 in.
ColorWhite
DisposableSingle-use
For Use With (Application)Simplify cleanup and safeguard work surfaces from stains, spills and wear
Quantity50 ft./Roll
Unit SizeEach