Glass Disposal Boxes
Glass Disposal Boxes
Actual product may vary
Glass Disposal Boxes
Separate broken and contaminated glassware from other waste.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelDimensions (L x W x H)
FISS41264Benchtop8 x 12 x 10 in.
FISS41263Floor12 x 12 x 27 in.
Catalog number FISS41264
Price (SGD)
-
Model:
Benchtop
Dimensions (L x W x H):
8 x 12 x 10 in.
  • When boxes are full of glass, simply remove the lid and flip up the rear flap to seal
  • Polybag liner for wet glassware can be sealed for safer disposal
  • Prevent injuries from broken glass
  • Printed for maximum visibility
  • Floor Model dimensions: 12L × 12W × 27 in.H (30.5 × 30.5 × 68.6cm)
  • Benchtop Model dimensions: 12L × 12W × 10 in.H (30.5 × 30.5 × 25.4cm)
Specifications
Dimensions (L x W x H)8 x 12 x 10 in.
Height (Metric)25.4 cm
Length (Metric)20.3 cm
ModelBenchtop
Quantity6/Cs.
Width (Metric)30.5 cm
Liner MaterialPolybag Liner
For Use With (Application)Separate broken and contaminated glassware from other waste
Width (English)12 in.
Length (English)8 in.
Height (English)10 in.
Unit SizeEA