Stainless Steel Laboratory Spoon Spatula
Stainless Steel Laboratory Spoon Spatula
Actual product may vary
Stainless Steel Laboratory Spoon Spatula
Stainless Steel Laboratory Spoon Spatula
Stainless Steel Laboratory Spoon Spatula
Stainless Steel Laboratory Spoon Spatula
Stainless Steel Laboratory Spoon Spatula
Stainless Steel Laboratory Spoon Spatula
Designed for use in professional laboratories, but is also perfect for home, classroom, kitchen & garage use.
Have Questions?
Catalog NumberLength (English)
FISS411496 in.
Catalog number FISS41149
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
6 in.
  • Dual Ended: 1 Flat End and 1 Spoon End
  • Designed for use in professional laboratories
  • Also suitable for use in Science Classrooms and Households
  • Chemical Resistant
  • Autoclavable
Specifications
Quantity1
AutoclavableAutoclavable
ShapeFlat Squared End, Spoon End
ColorSilver
Product TypeFlat End/Spoon Ended Spatula
For Use With (Application)Measuring
Length (English)6 in.
Width (Metric)0.25 cm
MaterialStainless Steel
Length (Metric)15 cm
Unit SizeEA