Disposable Polystyrene Weighing Dishes
Disposable Polystyrene Weighing Dishes
Actual product may vary
Disposable Polystyrene Weighing Dishes
Easy-grip contour sides for nonslip handling.
Have Questions?
Catalog NumberHeight (Metric)Length (Metric)Width (Metric)
FISS402921.9 cm8.3 cm8.3 cm
Catalog number FISS40292
Price (SGD)
-
Height (Metric):
1.9 cm
Length (Metric):
8.3 cm
Width (Metric):
8.3 cm
Perfect for staining or destaining electrophoresis mini-gels.
  • A pouring spout can be formed by squeezing
  • Shape ensures its flexibility on a balance platform
  • Smooth surface and rounded corners make powdered or granular samples easy to remove
Specifications
DisposableSingle-use
MaterialPolystyrene
ShapeSquare
TypeWeighing Dish
For Use With (Application)Perfect for staining or destaining electrophoresis mini-gels
Dimensions (L x W x H)3.3 x 3.3 x 0.75 in. (8.3 x 8.3 x 1.9 cm)
Length (English)3.3 in.
Length (Metric)8.3 cm
Width (English)3.3 in.
Width (Metric)8.3 cm
Height (Metric)1.9 cm
Height (English)0.75 in.
Unit SizeCS