Low Nitrogen Weighing Papers
Low Nitrogen Weighing Papers
Actual product may vary
Raylabcon
Low Nitrogen Weighing Papers
Sturdy and nonabsorbent.
Have Questions?
Catalog NumberDimensions (L x W)
FISS4028713 x 3 in. (76 x 76mm)
Catalog number FISS402871
Price (SGD)
-
Dimensions (L x W):
3 x 3 in. (76 x 76mm)
Sturdy and nonabsorbent.
  • Supports sample
  • Smooth texture avoids trapping of material
  • Will not absorb moisture
  • Sold in packs of 500
  • Packaged in 4 mil poly bag for extra protection
Specifications
Dimensions (L x W)3 x 3 in. (76 x 76mm)
For Use With (Application)For analytical balances
Length (English)3 in.
Length (Metric)76 mm
Width (English)3 in.
Width (Metric)76 mm
Product TypeLow Nitrogen Paper
ShapeSquare
Unit SizePack