Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Education Supplies
▸
Biotechnology Classroom Products
▸
Drosophila Genetics Biology Kits
▸
Murray the Mayfly, Blast Off! Deluxe Kit Components
Actual product may vary
Aqua Phoenix
Murray the Mayfly, Blast Off! Deluxe Kit Components
Replacement parts for Educational Kit
Have Questions?
Catalog Number
Product Type
FISS39691MKC
Size 64 rubber bands
Catalog number
FISS39691MKC
Price (SGD)
-
Product Type:
Size 64 rubber bands
Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.
Specifications
Product Type
Size 64 rubber bands
Grade
K to 5
Dimensions (L x W)
3.5 x 0.25 in.
For Use With (Application)
Education Kit
Quantity
0.25 lb. Bag