Check Order Status
Round Form Porcelain Evaporating Dishes
Actual product may vary
Fisherbrand
Round Form Porcelain Evaporating Dishes
Round Form Porcelain Evaporating Dishes are autoclavable and withstand temperatures up to 1150°C
Have Questions?
Catalog Number
Capacity (Metric)
Diameter (Metric)
FISS39335
80 mL
80 mm
Catalog number
FISS39335
Price (SGD)
-
Capacity (Metric):
80 mL
Diameter (Metric):
80 mm
Round form evaporating dishes include lip
Glazed inside and outside on both sides, except for large dishes (greater than 385 mL) which are glazed inside and partially outside
Specifications
Autoclavable
Autoclavable
Bottom Shape
Round
Capacity (Metric)
80 mL
Color
White
Material
Porcelain
Shape
Round
Sterility
Non-sterile
Surface Treatment
Glaze
Type
Evaporating Dish
Volume (Metric) Working
80 mL
Height (Metric)
30 mm
Diameter (Metric)
80 mm