Check Order Status
1.8mL Cryogenic Vials
Actual product may vary
Fisherbrand
1.8mL Cryogenic Vials
GSC 1.8mL Cryogenic Vials are used for storing biological samples at extreme temperatures as low as --196° Celsius.
Have Questions?
Catalog Number
Color
FISS39021
Clear
Catalog number
FISS39021
Price (SGD)
-
Color:
Clear
Specifications
Closure Color
Blue
Color
Clear
Material
Polypropylene
Volume (Metric)
1.8 mL
Height (English)
1.8 in.
No. per Pack
500
Style
Flat
Autoclavable
Not Autoclavable
Closure Type
Silicon Rubber-lined Cap
Height (Metric)
45 mm
Quantity
1