Polyethylene Safety Bottle Carriers
Polyethylene Safety Bottle Carriers
Actual product may vary
Polyethylene Safety Bottle Carriers
Carry up to eight bottles.
Have Questions?
Catalog NumberHolds
FISS37068Bottle (2 L)
Catalog number FISS37068
Price (SGD)
-
Holds:
Bottle (2 L)
  • Safe, simple way to carry bottles
  • Made of HDPE
  • One inch diameter carrying handle
Specifications
Product TypeBottle Carrier
AutoclavableNot Autoclavable
ColorWhite
HoldsBottle (2 L)
MaterialHDPE
Positions2
StackableNo
Dimensions (L x W x H)14.5 x 7.5 x 6.25 in. (36.8 x 19.1 x 15.9 cm)
Length (Metric)36.8 cm
Length (English)14.5 in.
Width (Metric)19.1 cm
Width (English)7.5 in.
Height (Metric)15.9 cm
Height (English)6.25 in.
Unit SizeEA