Polyethylene Safety Bottle Carriers
Polyethylene Safety Bottle Carriers
Actual product may vary
Polyethylene Safety Bottle Carriers
Carry up to eight bottles.
Have Questions?
Catalog NumberDescription
FISS37066Unpartitioned
Catalog number FISS37066
Price (SGD)
-
Description:
Unpartitioned
  • Safe, simple way to carry bottles
  • Made of heavy-duty HDPE
  • One inch diameter carrying handle
Specifications
Product TypeBottle Carrier
DescriptionUnpartitioned
AutoclavableNot Autoclavable
ColorWhite
MaterialHDPE
Positions1
StackableNo
Dimensions (L x W x H)14.5 x 7.5 x 6.25 in. (36.8 x 19.1 x 15.24 cm)
Length (Metric)36.8 cm
Length (English)14.5 in.
Width (Metric)19.1 cm
Width (English)7.5 in.
Height (Metric)15.24 cm
Height (English)6.25 in.
Unit SizeEA