Mortar and Pestle Sets
Mortar and Pestle Sets
Actual product may vary
Fisherbrand
Mortar and Pestle Sets
Porcelain mortar with pouring lip.
Have Questions?
Catalog number FISS337631
Price (SGD)
-
  • Glazed outside; rough inside
  • Pestles have glazed handles and unglazed ends
Specifications
Capacity (Metric)300 mL
FinishGlazed outside (mortar), glazed handle (Pestle)
Capacity (English)10.14 oz.
Diameter (English) Outer5.11 in.
Diameter (Metric) Outer13cm
Product TypeMortar and Pestle Set
MaterialPorcelain
Capacity10.14 oz., 300mL
Unit SizeEA