Marking Pens
Marking Pens
Actual product may vary
Marking Pens
Quick-drying ink has no unpleasant odor.
Have Questions?
Catalog NumberColor
FISS32179Black
Catalog number FISS32179
Price (SGD)
-
Color:
Black
  • Fast drying—highly resistant to abrasion and water
  • Designed to write on cold and damp surfaces as easily as they write on dry surfaces
  • Will not smear when subjected to water or alcohol during normal use
  • Ink is permanent on plastic, wood, and ceramic
  • Washes off glass, metal, or porcelain surfaces
Specifications
Product TypeLab Permanent Marker
Tip StyleFine
ColorBlack
SterilityNon-sterile
Latex FreeYes
For Use With (Application)Used to make permanent markings on plastic, metal, or porcelain labware
Quantity10 Pk.
Unit SizePK