Kartell™ Pipet/Buret Washing System Parts
Kartell™ Pipet/Buret Washing System Parts
Actual product may vary
Dynalon
Kartell™ Pipet/Buret Washing System Parts
Parts for Kartell™ Automatic Pipet Washing Set.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English)Height (English)Product Type
FISS3188016.5 in.25.5 in.Automatic Pipette Jar
FISS3188025 in.26 in.Pipet Basket
Catalog number FISS318801
Price (SGD)
-
Diameter (English):
6.5 in.
Height (English):
25.5 in.
Product Type:
Automatic Pipette Jar
Two or three baskets can be used with one or two jars and one rinser for economy of operation. Since the soaking operation takes the longest, two or three batches of glassware can be soaking at the same time.
  • Hold pipets or burets up to 24 in. (60.8cm) long
  • Polyethylene construction reduces breakage and buffers the tops of pipets against the bottom of jars
  • Back-siphoning is eliminated because water inlet is located at top of rinser
  • Diameter of tubing permits water to drain completely
  • Rinser repeats automatically
For complete unit, see S31880A. S31880 is washer unit only and requires S318801 and S318802. Tubing not included.
Specifications
Height (English)25.5 in.
Height (Metric)64.77 cm
Diameter (English)6.5 in.
Diameter (Metric)16.5cm
MaterialPolyethylene
For Use With (Equipment)For pipet/buret washing system
Product TypeAutomatic Pipette Jar
Unit SizeEach