Check Order Status
Kimble™ Amber Glass Dropping Bottles
Actual product may vary
DWK Life Sciences
Kimble™ Amber Glass Dropping Bottles
For storage of light-sensitive chemicals.
Have Questions?
Catalog Number
Capacity (Metric)
FISS31683
30 mL
Catalog number
FISS31683
Price (SGD)
-
Capacity (Metric):
30 mL
Polypropylene screw cap with rubber bulb
Glass dropper inside
Choose from 30 or 60mL capacity
Specifications
Capacity
1 oz., 30mL
Capacity (Metric)
30 mL
Capacity (English)
1 oz.
Color
Amber
Material
Glass
Closure Material
Polypropylene
Closure Type
Screw Cap