Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap
Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap
Actual product may vary
Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap
General purpose bottle. Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap are used for pharmaceutical, chemical, environmental, and food packaging and storing applications.
Have Questions?
Catalog NumberCapacity (Metric)
FISS31666-F120 mL
Catalog number FISS31666-F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
120 mL
Clear / Flint Standard Wide Mouth Bottles feature a wide opening making them the ideal general purpose bottle. Applications include pharmaceutical, chemical, environmental, and food packaging and storing.
  • Made of flint glass
  • Lids are flat, disc-type and polyvinyl-lined black phenolic
  • Wide opening, ideal for general purpose
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
Specifications
Capacity4 oz., 120mL
Capacity (Metric)120 mL
Capacity (English)4 oz.
ColorClear
MaterialFlint Glass, Glass
Closure ColorBlack
Product TypeStandard Wide Mouth Bottle
Closure LinerPoly/Vinyl
Closure MaterialPhenolic
Closure TypeScrew Cap
Unit SizeCS