Kimble™ KIMAX™ Beaker, Griffin, Low Form
Kimble™ KIMAX™ Beaker, Griffin, Low Form
Actual product may vary
Kimble™ KIMAX™ Beaker, Griffin, Low Form
Kimble™ KIMAX™ Beaker, Griffin, Low Form
DWK Life Sciences
Kimble™ KIMAX™ Beaker, Griffin, Low Form
Thick-walled beakers for a standard of durability in the classroom
Have Questions?
Catalog NumberCapacity (Metric)
FISS30730-8600 mL
Catalog number FISS30730-8
Price (SGD)
-
Capacity (Metric):
600 mL
  • Beaded top, with spout designed to prevent dripping
  • Durable borosilicate glass beakers
  • Graduated scales
  • Large, scratch- and chemical-resistant marking spot
Specifications
Capacity (English)20.28 oz.
Diameter (English) Outer3.46 in.
Diameter (Metric) Outer8.8 cm
Capacity (Metric)600 mL
MaterialBorosilicate Glass
For Use With (Application)Excellent Mechanical Strength and Durability, While Providing High Resistance to Chemical Attack and Thermal Shock
Graduation Range50 to 500 mL
Height (English)4.8 in.
Height (Metric)12.2 cm
IncludesSpout
Increments50 mL
Label TypeMatte Finish Marking
Quantity6 Pk.
TypeBeaker
GraduatedYes
Certifications/ComplianceASTM E960, ASTM E438, USP Type I
ScaleDual Scale
StyleGriffin
Unit SizePK