Check Order Status
Flexible Brushes
Actual product may vary
Flexible Brushes
Soft, single row of bristles fits easily into flask necks.
Have Questions?
Change view
Catalog Number
Length (English)
For Use With (Equipment)
FISS30410C
16.9 in.
1000mL flasks
FIS03-570B
14.17 in.
500mL flasks
2 Options
Catalog number
FISS30410C
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
16.9 in.
For Use With (Equipment):
1000mL flasks
Soft, single row of bristles fits easily into flask necks.
Plastic handle bends to allow bristles to fit into container
Handle has hanging ring
Specifications
Product Type
Flexible Brush
Bristle Type
Soft
Handle Material
Plastic
Handle Type
Flexible
Diameter (English) Bristle
1.7 in.
Diameter (Metric) Bristle
4.4 cm
For Use With (Equipment)
1000mL flasks
Includes
Hanging ring
Length (English)
16.9 in.
Length (English) Bristle
5.11 in.
Length (Metric)
43 cm
Length (Metric) Bristle
13 cm
No. of Bristle Rows
1