Steritop™ Threaded Bottle Top Filters
Steritop™ Threaded Bottle Top Filters
Actual product may vary
MilliporeSigma
Steritop™ Threaded Bottle Top Filters
The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.
Have Questions?
Catalog NumberVolume (Metric)Material (Membrane)
FISS2GPT05RE500 mLPES
Catalog number FISS2GPT05RE
Price (SGD)
454.00
CS
Add to cart
Volume (Metric):
500 mL
Material (Membrane):
PES
Price (SGD)
454.00
CS
Add to cart
The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.
  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
  • Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
  • Sterilization of Aqueous Solutions (150 mL to 1 L)
    • Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
Specifications
ColorClear with blue adapter
DisposableSingle-use
Filter TypeMembrane Filter
MaterialPolystyrene
Material (Membrane)PES
Pore Size0.22 μm
SterilitySterile
For Use With (Application)Sterile filtration
Outlet Fitting45 mm
QuantityPack of 12
TypeBottle Top Filter
Volume (Metric)500 mL
Length (Metric)263 mm
Height (Metric)148 mm
Diameter (Metric)12.5 cm
Diameter (Metric) Filter12.5 cm
Diameter (Metric) Membrane12.5 cm
Unit SizeCS