Finger Print Classification STEM Kit
Finger Print Classification STEM Kit
Actual product may vary
Finger Print Classification STEM Kit
Innovating Science™ Finger Print Classification STEM Kit allows students to learn about the history of fingerprinting as a tool for identification.
Have Questions?
Catalog NumberClass Size
FISS2955915 Lab Groups
Catalog number FISS29559
Price (SGD)
-
Class Size:
15 Lab Groups
  • Analyze data using the system that is used worldwide for classification
  • Analyze fingerprints for common characteristics and create a classification system using those characteristics
  • This kit includes an optional activity where students can practice lifting a fingerprint and comparing it to a crime scene print
  • Kit contains enough materials for 15 groups
  • Teacher's guide and Student Guide copymasters are included
Specifications
Product TypeSTEM Kit
ColorWhite
Grade7 to 14
Age13 to 20
For Use With (Application)Forensics Class (for groups of 15)
IncludesTeachers Manual and Student Guide
Quantity1 Kit
Dimensions (L x W x H)9 x 6.5 x 2.75 in.
Class Size15 Lab Groups
Unit SizeEA