Hollow Glass Hexagonal Stoppers
Hollow Glass Hexagonal Stoppers
Actual product may vary
Hollow Glass Hexagonal Stoppers
Feature a round end and come in a variety of join sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStandard Taper Joint
FISS2949724/29
FISS2949514/23
FISS2949619/26
Catalog number FISS29497
Price (SGD)
-
Standard Taper Joint:
24/29
  • Hollow glass stopper
  • Features a round end with ground glass for a snug fit
  • Head has a hexagonal shape
  • Used to seal glass flasks and bottles
  • Made from high quality borosilicate 3.3 glass
  • Autoclavable
  • 1 year limited manufacturer warranty
    Specifications
    DescriptionGlass Stoppers
    Diameter (Metric)2.4 cm
    For Use With (Application)Seal flasks
    Height (Metric)3.6 cm
    Length (Metric)2.4 cm
    Quantity1
    Size24/29
    Standard Taper Joint24/29
    Width (Metric)2.4 cm
    AutoclavableAutoclavable
    ColorClear
    Diameter (Metric) Outer24 mm
    MaterialBorosilicate Glass
    Product TypePennyhead Stopper
    ShapeCylindrical
    Unit SizeEA