Nylon Flask Brush
Nylon Flask Brush
Actual product may vary
Nylon Flask Brush
Nylon Flask Brush
Nylon Flask Brush
Nylon Flask Brush
Fisherbrand
Nylon Flask Brush
Nylon Flask Brush is comprised of durable nylon bristles and a stainless steel wire handle.
Have Questions?
Catalog NumberColor
FISS29383White
Catalog number FISS29383
Price (SGD)
-
Color:
White
  • Durable nylon bristles
  • Stainless steel wire handle
  • Curved design for thorough cleaning of round bottom glassware
Specifications
Product TypeCleaning Brush
Bristle MaterialNylon
Bristle TypeMedium/Stiff
Color-codedNo
Handle MaterialStainless Steel Wire
Handle TypeTwisted Wire
Tip TypeFan Tip
ScentUnscented
SterilityNon-sterile
ColorWhite
For Use With (Application)Clean labware
For Use With (Equipment)Glassware, plasticware
Length (English)13 in.
Length (Metric)35 cm
Quantity1 Ea.
Unit SizeEA