Plastic Dropping Bottle, Euro Design
Plastic Dropping Bottle, Euro Design
Actual product may vary
Fisherbrand
Plastic Dropping Bottle, Euro Design
Unique LDPE Plastic Dropping Bottles include a screw cap and spout that dispenses small, uniform drops of liquid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FISS29378125 mL1 Ea.
FISS2937515 mL1 Ea.
FISS2937760 mL1 Ea.
Catalog number FISS29378
Price (SGD)
-
Capacity (Metric):
125 mL
Quantity:
1 Ea.
  • Plastic Dropping Bottle
  • Low Density Polyethylene (LDPE)
  • Spout dispenses small, uniform drops of liquid
  • Screw cap with an attached cover
  • Euro design
Specifications
Capacity (Metric)125 mL
Capacity (English)4.2 oz.
ColorClear
DescriptionUniform drops
Diameter (English)2 in.
For Use With (Application)Dispensing liquids
IncludesScrew cap and spout
Height (Metric) With Closure50.8 mm
MaterialLDPE
Length (Metric)6.3 cm
Quantity1 Ea.
SterilityNon-sterile
Width (Metric)6.3 cm
Closure ColorTranslucent
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
Drop SizeSmall
GraduatedNo
MouthNarrow
ShapeCylindrical
Height (Metric)5 cm
Product TypeDropping Bottle
For Use With (Equipment)Liquids
Width (English)2 in.
Height (English)2 in.
Closure MaterialPlastic
Diameter (Metric)6.3 cm
Closure TypeScrew Cap
Unit SizeEA