Reusable Polyethylene Dippers
Reusable Polyethylene Dippers
Actual product may vary
Reusable Polyethylene Dippers
500 mL beakers with long handles for removing samples from large tanks.
Have Questions?
Catalog NumberLength (Metric) HandleLength (English) Handle
FISS290770.9 m3 ft.
Catalog number FISS29077
Price (SGD)
-
Length (Metric) Handle:
0.9 m
Length (English) Handle:
3 ft.
  • Lightweight, durable polyethylene
  • Graduated beaker features dual pouring spouts for easy left- or right-handed operation
  • Handle has an “easy-grip” end for secure handling under adverse conditions
Specifications
Product TypeDipper
MaterialHDPE
Capacity (Metric)500 mL
For Use With (Application)For removing samples from large tanks
Dimensions (L x W x H)5.75 x 5 x 43 in. (14.6 x 12.7 x 109.22 cm)
Height (English)43 in.
Height (Metric)109.22 cm
Length (English)5.75 in.
Length (English) Handle3 ft.
Length (Metric)14.6 cm
Length (Metric) Handle0.9 m
Width (English)5 in.
Width (Metric)12.7 cm
Unit SizeEA