mySPIN™ Mini Centrifuge Series
mySPIN™ Mini Centrifuge Series
Actual product may vary
Fisherbrand
mySPIN™ Mini Centrifuge Series
Bring increased efficiency and user comfort to your lab with Thermo Scientific mySPIN 6 and 12 mini centrifuges
Have Questions?
Catalog NumberCapacity
FISS2843512 x 2 mL
Catalog number FISS28435
Price (SGD)
-
Capacity:
12 x 2 mL
Tool-free rotors and quick-spin options make them fast and easy to use. Benefit from a compact footprint that fits into your workstation and ergnonomic design that stands out. Ideal for microfilter cell separation and HPLC protocols, with the mySPIN 12 centrifuge also supporting PCR, as well as clinical and diagnostic applications.
Benefit from:
  • Compact footprint
  • Easy operation
  • Low noise, low vibration
  • Tool-free, quick rotor exchange
mySPIN 12 mini centrifuge also features:
  • Imbalance and tilt safety cutoff, as well as safety interlock lid designed to prevent opening while rotor is turning
  • Flexibility: ideal for most protocols
  • Ability to perform fast spins with higher RCF requirements
  • Back-lit, customizable display that can be set for rpm or RCF
mySPIN 6 mini centrifuge also offers:
  • Safety features: unit will not operate without lid in place
  • Quick-spin feature
  • Palm-shaped lid for left- or right-handed users
  • Quick tube extraction
Applications:
  • mySPIN 12 centrifuge is ideal for most protocols requiring fast spins (12,500rpm/9,800 x g), and appropriate for PCR, microfilter cell separation, and HPLC protocols
  • mySPIN 6 is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2mL)
Specifications
Capacity12 x 2 mL
Max. Speed12,500 rpm
Max. RCF9,800 x g
DisplayDigital
Noise Level51-53 dB(A)
Certifications/ComplianceCE, CSA, UL
Includes12 x 1.5/2.0mL tube rotor, 32 x 0.2mL tube rotor (singles or 4 x 8 strip), 12 tube adapters 0.2mL, 12 tube adapters 0.5mL, universal power adapter 110-240V 50/60 Hz, with 4 interchangeable plug type
Electrical Requirements110/240 V 50/60 Hz with 4 interchangeable plug adapters
RefrigeratedNo
Run Time1 sec. to 30 min. (1 sec. increments) and Quick Spin available
Dimensions (HWD)4.4 x 6.7 x 8 in. (11.4 x 17.1 x 20.3 cm)
Net Weight3.1 lb. (1.41 kg)
Unit SizeEA