Check Order Status
Polyvinyl Alcohol Solution, 4%
Element Search
Structure Search
Polyvinyl Alcohol Solution, 4%
Have Questions?
Catalog Number
Quantity
Packaging
FISS25474
100 mL
Thick Gel - 4% Solution
Catalog number
FISS25474
Price (SGD)
-
Quantity:
100 mL
Packaging:
Thick Gel - 4% Solution
Request bulk or custom format
Specifications
Chemical Name or Material
Polyvinyl Alcohol
Shelf Life
2 years
CAS
9002-89-5
Physical Form
Solid
Molecular Formula
(C2H4O)n
View more