Check Order Status
Crystal Violet, 1%, Solution
Element Search
Structure Search
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Crystal Violet, 1%, Solution
Have Questions?
Contact Us
Catalog Number
Quantity
Packaging
FISS25275A
100 mL
1%
Catalog number
FISS25275A
Price (SGD)
-
Contact Us
Quantity:
100 mL
Packaging:
1%
Request bulk or custom format
Specifications
Chemical Name or Material
Crystal Violet, Solution
Shelf Life
18 months
Synonym
Gentian Violet, Gram Stain Solution
Packaging
1%
Density
1g/mL
View more
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link