Stainless Steel Clamp - 3 Prong Cork Lined Jaws - Holding Capacity 90mm
Stainless Steel Clamp - 3 Prong Cork Lined Jaws - Holding Capacity 90mm
Actual product may vary
Stainless Steel Clamp - 3 Prong Cork Lined Jaws - Holding Capacity 90mm
High quality, durable 3 prong clamp with cork lined steel jaw that holds objects up to a 100mm or 4 in. diameter . Clamp tightens securely using a convenient thumbscrew. Overall clamp length is 11 in. and the diameter of the rod is 8mm.
Have Questions?
Catalog NumberColor
FISS24278Silver
Catalog number FISS24278
Price (SGD)
-
Color:
Silver
Specifications
TypeAdjustable Clamp
MaterialZinc Die Cast, Plastic Coated
Height (Metric)2.54 cm
Height (English)1 in.
Length (English)11 in.
Length (Metric)27.94 cm
Width (English)4 in.
Width (Metric)10.16 cm
ColorSilver
Jaw TypeFixed
Unit SizeEA