Glass Erlenmeyer Flasks, Narrow Neck
Glass Erlenmeyer Flasks, Narrow Neck
Actual product may vary
Glass Erlenmeyer Flasks, Narrow Neck
Graduated conical flasks made from high-quality borosilicate 3.3 glass.
Have Questions?
Catalog NumberCapacity (Metric)
FISS23900250 mL
Catalog number FISS23900
Price (SGD)
-
Capacity (Metric):
250 mL
Borosilicate glass Erlenmeyer flasks. 250mL capacity.
  • High-quality borosilicate 3.3 glass
  • Feature beaded rims and narrow neck
  • White, chemical resistant graduations and markings
  • Low Temperature gradient - can withstand high temperatures and thermal shocks
  • Superior chemical durability
  • Autoclavable at 121°C
  • Eisco LabGlass has the following heat tolerances: Strain point -515°C, annealing point 565°C, softening point 820°C
Specifications
ColorClear
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Graduation Range100 to 250 mL
Neck StyleNarrow
Product TypeErlenmeyer Flask
Writing SurfaceYes
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Mouth StyleBeaded
SterilityNon-sterile
Bottom ShapeConical
Surface TreatmentNon-treated
Quantity12/Pk.
Unit SizeEA