Graduated Cylinder, Premium Polypropylene, Hexagonal Base - Class B
Graduated Cylinder, Premium Polypropylene, Hexagonal Base - Class B
Actual product may vary
Graduated Cylinder, Premium Polypropylene, Hexagonal Base - Class B
Graduated Cylinder, Premium Polypropylene, Hexagonal Base - Class B
Graduated Cylinder, Premium Polypropylene, Hexagonal Base - Class B
Polypropylene graduated cylinders have a hexagonal base, are made from a single piece and are autoclavable.
Have Questions?
Catalog NumberCapacity (Metric)
FISS2379610, 25, 50, 100, 250, 500, 1000 mL
Catalog number FISS23796
Price (SGD)
-
Capacity (Metric):
10, 25, 50, 100, 250, 500, 1000 mL
  • Hexagonal base
  • Made from a single piece
  • Autoclavable
Specifications
Capacity (Metric)10, 25, 50, 100, 250, 500, 1000 mL
MaterialPolypropylene
DescriptionSet of 7
CalibrationTo contain
DisposableReusable
ScaleRaised graduation
Quantity1
Graduations10 mL
AutoclavableAutoclavable
Capacity (English)0.38 to 33.8 oz.
ShapeRound
SterilityNon-sterile
Scale ColorClear
Base TypeHexagonal
Height (English)6 in.
Height (Metric)152.4 mm
TypeGraduated Cylinder Set
Unit SizePK