Nylon Test Tube Brush
Nylon Test Tube Brush
Actual product may vary
Nylon Test Tube Brush
Nylon Test Tube Brush
Nylon Test Tube Brush
Nylon Test Tube Brush
Nylon Test Tube Brush
With fan-shaped end.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorLength (Metric)Diameter (Metric) Bristle
FISS23787White228.6 mm29 mm
FISS13962White230 mm20 mm
Catalog number FISS23787
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
White
Length (Metric):
228.6 mm
Diameter (Metric) Bristle:
29 mm
  • Made from nylon
  • Wire-wound handle with loop
Specifications
Product TypeTest Tube Brush
Bristle MaterialNylon
Bristle TypeMedium Stiffness
Color-codedNo
Handle MaterialStainless Steel Wire
Handle TypeTwisted Wire
Tip TypeFan Tip
ScentUnscented
SterilityNon-sterile
ColorWhite
Diameter (English) Bristle1.125 in.
Diameter (Metric) Bristle29 mm
For Use With (Application)For cleaning test tubes
Length (English)9 in.
Length (English) Brush3.5 in.
Length (Metric)228.6 mm
Length (Metric) Bristle88.9 mm
Length (Metric) Brush88.9 mm
Quantity1 Ea.
Unit SizeEA