Food Nutrient Analysis Kit
Actual product may vary
Lab-Aids
Food Nutrient Analysis Kit
Test common foods for presence of nutrients, quantitatively determine Vitamin C in foods and standardize all tests.
Have Questions?
Catalog NumberClass Size
FISS1934030 Students
Catalog number FISS19340
Price (SGD)
-
Class Size:
30 Students
Working in teams, students are asked to test for, and identify, five classes of nutrients: sugar, starch, lipid, protein, and Vitamin C. Once the tests have been completed and their observations have been made, students are then asked to draw conclusions about their results, the controls, and samples that showed no reaction. It is necessary to have a hot water bath available for the test of simple sugars. This kit has a limited shelf-life. Includes Teacher guide, student sheets, assessment questions and SDS.
Specifications
For Use With (Application)Test and identify five classes of nutrients like sugar, starch, lipid, protein, and vitamin c in foods
IncludesTeacher's guide with MSDS, 30 Student Worksheets and Guides, Drop control bottle of Glucose, Vitamin C, Protein, Starch Suspension, Vegetable Oil, 2 Fat Testing Solution, 2 Protein Testing Solution A and B, 2 Sugar Testing Solution, 2 Vitamin C Testing Solution, 2 Bottles of Starch Testing solution, 30 Test tubes
Time RangeOne ∽50 minute class period
Class Size30 Students
Product TypeFood Nutrient Analysis Kit
Unit SizeEach