Check Order Status
Disposable Cover Slips
Actual product may vary
Disposable Cover Slips
Unbreakable, noncurling, plastic slips.
Have Questions?
Change view
Catalog Number
Quantity
FISS17525C
1000/Pk.
FISS17525B
100 Pk.
2 Options
Catalog number
FISS17525C
Price (SGD)
-
Quantity:
1000/Pk.
Unbreakable, noncurling, plastic slips.
Size: 0.87 × 0.87 in. (22 × 22mm)
Thickness: 0.007 × 0.010 in. (0.17 to 0.25mm)
Specifications
Product Type
Coverslip
Material
Plastic
Length (Metric)
22 mm
Width (Metric)
22 mm
Thickness (Metric)
0.17 to 0.25 mm
Disposable
Single-use
Quantity
1000/Pk.